Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

29546-U

Supelco

High Capacity Gas Purifier, Starter Kit

AC/DC input 230 V AC, size 1/8 in., CE Approved

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813
NACRES:
SB.54

size

1/8 in.

Looking for similar products? Visit Product Comparison Guide

General description

Conformity Marks: CE Approved
No other purifier removes both moisture and oxygen in such large quantities (35 liters of moisture, 14 liters of oxygen ).

Protects GC columns and detectors from damage caused by moisture and oxygen in the carrier gas stream, even when present up to 100 ppm levels. The clam-shell oven heats a converter tube to 580°C, causing moisture and oxygen to irreversibly react with the reactant material. This chemical reaction prevents contaminants from returning to the gas stream, even when the material approaches saturation, or when the oven is cooled. Carbon monoxide and carbon dioxide are also removed.

It is recommended to install 4 ft. (1.2 m) of tubing downstream of the oven to allow heat to dissipate from the gas stream. This tubing can be coiled to reduce space.

The starter kit includes a clam-shell oven, two elements (installed), and a converter tube.

Note: Not for use with hydrogen gas.

Clam-Shell Oven:
  • Length: 14 1/2 in. (36.8 cm)
  • Height: 5 1/4 in. (13.3 cm)
  • Depth: 6 in. (15.2 cm)
  • Operating Temp.: 580°C
  • Power Consumption: 90 watt
  • Conformity marks: CE Approved
  • Mounting: Horizontally only (bench or wall) using integral bracket
  • Warranty: 1 year

Converter Tube:
  • Length inside column oven: 10 in. (25.4 cm)
  • Diameter: 1/2 in. (12.7 mm)
  • Max. Inlet Pressure: 150 psi
  • Max. Flow Rate: 1100 mL/min

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service