29329-U
Center Drain (CD) screw thread vials, vial only
volume 3.8 mL, amber glass vial, O.D. × H 14.75 mm × 45 mm, thread for 13-425, pkg of 100 ea
About This Item
material
amber glass vial
feature
cap is bonded to septa to prevent fallout
packaging
pkg of 100 ea
O.D. × H
14.75 mm × 45 mm
volume
3.8 mL
fitting
thread for 13-425
Looking for similar products? Visit Product Comparison Guide
General description
- Available in 3.8 to 60 mL size.
- Less than 4 μL dead volume.
- The sturdy CD Vial with the patented unique is ideal for many micro applications.
- The vials are manufactured from Boroisilicate Type 1 glass to safeguard against a change in the pH of the contents.
The 12 x 32 mm CD Vial is available in snap top, crimp top, and 9 mm screw thread finish. This vial offers a 1.5 ml sample volume, works in all autosamplers, provides maximum content retrieval without requiring a limited volume insert. The production process provides consistency of quality and precision throughout all sizes of CD Vials. Bottom thickness dimension is the same in each vial and the conical well is located in the exact center of each vial. The patented CD Vial incorporates a smooth transition area from the sidewall of the vial to the conical well. This allows the contents of the vial to drain to the center of the conical well for maximum extraction (< 4ul dead volume).
Application
- Library sample storage
- Lyophilize
- Centrifugation
- Micro mixing
- Extraction
- Concentrations
- Derivatization
- Small scale reactions
Other Notes
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service