Skip to Content
MilliporeSigma
All Photos(2)

Documents

28714-U

Supelco

Empty Tube for Thermal Desorption

greener alternative

glass, O.D. x I.D. x L 1/4 in. × 3 1/2 in., non-fritted, pkg of 10 tubes

Synonym(s):

empty glass Thermal Desorption Tube (unfritted)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

product name

Empty Tube for Thermal Desorption, glass TD tube (non-fritted), O.D. × L 1/4 in. × 3 1/2 in., with (Tension Springs (qty 10)), pkg of 10 tubes

material

glass TD tube (non-fritted)

packaging

pkg of 10 tubes

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2 in.

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Empty tube for thermal desorption is a glass thermal desorption tube, used for packing other adsorbents or sample preparation applications.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

related product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service