28521-U
ATIS Adsorbent Tube Injector System 230V
Synonym(s):
ATIS Injector System
About This Item
Recommended Products
application(s)
air monitoring
environmental
industrial hygiene
Related Categories
General description
Features and Benefits
- Injecting calibration standards onto adsorbent tubes, to calibrate your analytical system
- Injecting surrogates and system monitoring compounds onto adsorbent tubes before or after sampling
- Removes moisture from tubes prior to analysis (Dry purging)
- Connect an air-sampling bag to the outlet of the ATIS to vaporize a calibration standards prior to assure complete
The ATIS will accept either 1/4in. or 6mm O.D. Thermal Desorption tubes. Included is a Luer/Hose Barb adapters to connect a variety of solvent desorption tubes. The temperature range of the ATIS is ambient to 120°C. The
flow range is 0 to 100mL/min. The ATIS includes the injection glassware, a constant flow controller with a on/off valve, the heating source, spare parts along with all the necessary fittings and tubing. You simply plumb it to a regulated source of nitrogen or helium and plug it in to the appropriate electrical source.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Discover our new Carbotrap® T420 tube, designed for precise terpene air sampling in the cannabis and hemp industry. Monitor terpenes in cultivation environments, even in high humidity, with reproducible sample recovery. Elevate your analysis with the latest in terpene sampling technology.
Chromatograms
suitable for GC, application for air monitoringsuitable for GC, application for air monitoringsuitable for GC, application for air monitoringsuitable for GC, application for air monitoringShow MoreOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service