Key Documents
28408-U
Supelco
Vacuum Adapter
I.D. 10 mm, pk of 1
About This Item
UNSPSC Code:
13111000
NACRES:
NB.21
Pricing and availability is not currently available.
description
for procesing packed and empty dioxin columns and dual-layer reversible carbon tubes individually using vacuum pressure
packaging
pk of 1
technique(s)
solid phase extraction (SPE): suitable
I.D.
10 mm
application(s)
food and beverages
Choose from one of the most recent versions:
Certificates of Analysis (COA)