Key Documents
28405-U
Supelco
Syringe Luer Adapter
I.D. 10 mm, pk of 3
Select a Size
3 EA
$162.00
$162.00
Please contact Customer Service for Availability
Request a Bulk OrderRequest a Sample
Select a Size
Change View
3 EA
$162.00
About This Item
UNSPSC Code:
13111000
NACRES:
NB.21
$162.00
Please contact Customer Service for Availability
Request a Bulk OrderRequest a Sample
description
for processing Dual-Layer Carbon Reversible Tubes using a glass luer syringe
packaging
pk of 3
technique(s)
solid phase extraction (SPE): suitable
I.D.
10 mm
application(s)
food and beverages
Choose from one of the most recent versions:
Certificates of Analysis (COA)