Key Documents
2630901
Supelco
Inlet Liner, Splitless Type, Straight Design (unpacked)
pkg of 1 ea
About This Item
UNSPSC Code:
12000000
packaging
pkg of 1 ea
L × O.D. × I.D.
92 mm × 6.35 mm × 4.0 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits PerkinElmer (AutoSystem)
Choose from one of the most recent versions:
Certificates of Analysis (COA)