Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24865

Supelco

PAS-500 Micro Air Sampler with Low Flow Orifice

Includes 6 mm tube holder, screwdriver, and two 9-volt batteries

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008

feature

Includes 6 mm tube holder, screwdriver, and two 9-volt batteries

packaging

pkg of 1 ea

technique(s)

active air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

General description

This low flow pump is lightweight (4 oz.) and compact (7 in. H) with a flame-retardant case, fitting easily into your shirt pocket. The adsorbent tube connects directly to the inlet of the pump. The PAS-500 sampler is versatile and adapts to fit both 6 mm and 8 mm diameter tubes. The flow range is 5-200 cc/min. An optional low flow adapter enables you to sample at 20 cc/min.

The unit is powered by a convenient, easily replaceable 9-volt battery. A full flow regulation feature provides constant voltage to the pump, even as the battery voltage drops. This sampler is intrinsically safe but not suitable for sampling in explosive environments (NOT EX-Rated). The design features a current-limiting resistor in the battery circuit which prevents danger of short-circuit spark. CE Approved.

recommended

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service