Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24647

Supelco

Vacuum Sampling Pumps

Model 1063 Bag Sampler (Six 1 Liter Bags)

Synonym(s):

Air Sampling Pumps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008

description

Not UL listed; Not EX rated

technique(s)

whole air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with Six 1 Liter Bags

Looking for similar products? Visit Product Comparison Guide

General description

Vacuum Sampling Pumps is a field sampling pump, used as accessories for active and whole air samplers such as Model 1063 bag sampler.

Other Notes

A Critical Orifice can be ordered as part number 24667 if required as a replacement item. The filter for the critical orifice is part number 24672.
The battery charger is not included with part number 24647. Please order part number 24644 separately.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service