Skip to Content
MilliporeSigma
All Photos(1)

Documents

24635

Supelco

Battery for Vacuum Sampling Pump

for use with Model 1060

Synonym(s):

Battery for Field Sampling Pump

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008

technique(s)

whole air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with Model 1060

Looking for similar products? Visit Product Comparison Guide

General description

Battery for Vacuum Sampling Pump are accessories for field sampling pumps. It is a DC battery employed with a 1-liter vacuum bag sampler.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service