Key Documents
23783
Needle File
4 in. long, slim tapered files, pkg of 2 ea
About This Item
UNSPSC Code:
41100000
Pricing and availability is not currently available.
packaging
pkg of 2 ea
General description
4 in. long, slim taper files.
Use to enlarge ferrule I.D. Also handy for removing ferrules stuck in fittings.
Choose from one of the most recent versions:
Certificates of Analysis (COA)