Documents
23740-U
Supelco
Capillary Cleaving Tool
Fixed Blade Version
About This Item
UNSPSC Code:
12000000
NACRES:
SB.54
material
sapphire cutting blade
packaging
pkg of 1 ea
General description
This handy tool makes scalpel-like cuts to polyimide-coated fused silica, leaving no jagged edges to create problems. The industrial sapphire cutting edges remains sharp indefinitely.
Other Notes
Fixed blade with a protective cover.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)