Documents
23240-U
Supelco
Screw cap
black phenolic solid cap (with polyseal liner), thread for 22-400, pkg of 144 ea
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
black phenolic solid cap (with polyseal liner)
packaging
pkg of 144 ea
fitting
thread for 22-400
compatibility
for use with 120 mL (49 × 114 mm)
General description
Screw caps (thread 22-400) are available for use with sealing septa and solid caps with polyseal liners. Caps are offered in solid phenolic caps that can withstand a maximum temperature of 205° C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)