Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23017

Supelco

HPLC sample loop 10μl ss 7000psi ea

sample loop volume 10 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

material

stainless steel sample loop and nuts

manufacturer/tradename

VICI Valco® SL10CUW

parameter

7000 psi max. pressure

sample loop volume

10 μL

fitting

10-32 Valco fittings (nuts and ferrules included)

compatibility

for use with Valco HPLC valve

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Valco is a registered trademark of Valco Instruments Co. Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuma Kumamoto et al.
Neuron, 107(4), 617-630 (2020-06-20)
Stable genomic integration of exogenous transgenes is essential in neurodevelopmental and stem cell studies. Despite tools driving increasingly efficient genomic insertion with DNA vectors, transgenesis remains fundamentally hindered by the impossibility of distinguishing integrated from episomal transgenes. Here, we introduce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service