Key Documents
22948
Supelco
Zero Volume Reducer 1/16 to 1/8 External
About This Item
UNSPSC Code:
41115700
material
stainless steel
packaging
pkg of 1 ea
fitting
1/16 in. Valco (nut and ferrule included)
1/8 in. male Valco for 1/8 in. injector or detector
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)