Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22823-U

Supelco

Supelpure HC Hydrocarbon Trap Refill

volume 474 cc

Sign Into View Organizational & Contract Pricing

Select a Size

474 CC
$96.70

$96.70


Available to ship onApril 11, 2025Details


Request a Bulk Order

Select a Size

Change View
474 CC
$96.70

About This Item

EC Number:
UNSPSC Code:
12000000
NACRES:
SB.54

$96.70


Available to ship onApril 11, 2025Details


Request a Bulk Order

volume

474 cc

Looking for similar products? Visit Product Comparison Guide

General description

Activated charcoal.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service