Skip to Content
MilliporeSigma
All Photos(1)

Documents

22775

Agilent/HP JetPaper Print Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
44103103

General description

Fits the Agilent/HP 3396A integrator, plus QuietJet, ThinkJet, and QuietJet Plus printers. One cartridge prints ~600,000 characters in black ink.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

437.0 °F

Flash Point(C)

225 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service