Documents
22509
Supelco
Supelco Ferrule Nut Adapter (fits Agilent injectors and non-MS detectors), knurled fingertight version
pkg of 2 ea
About This Item
UNSPSC Code:
41121815
packaging
pkg of 2 ea
General description
These stainless steel adapters enable you to use general purpose and other 1/16 in. compression ferrules in Agilent GCs. Knurled head allows tightening with fingers, eliminating the need to locate the proper wrench.
Choose from one of the most recent versions:
Certificates of Analysis (COA)