Skip to Content
MilliporeSigma
All Photos(1)

Documents

22086-U

Supelco

Supeltex M-1 Packed Column Ferrule, 1/4 in. Column O.D.

I.D. 1/4 in., 1/4 in. O.D. Column, pkg of 10 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
SB.54

feature

ferrule type packed column

packaging

pkg of 10 ea

I.D.

1/4 in.

compatibility

1/4 in. O.D. Column

Looking for similar products? Visit Product Comparison Guide

General description

Characteristics: Ideal for connection to mass spectrometers. High reusability. Isothermal use only.

Material Composition: Ceramic-filled PTFE

Maximum Temperature:
250 °C

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service