22042-U
Swagelok® Reducing Union
Swagelok® 200-6-1, stainless steel, 1/16 in. Swagelok, 1/8 in. Swagelok
Synonym(s):
Swagelok® 200-6-1,SS REDUCING UNION 1/8X1/16 EA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41112500
eCl@ss:
32035302
NACRES:
SB.54
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
Swagelok® 200-6-1
fitting
1/16 in. Swagelok
1/8 in. Swagelok
Looking for similar products? Visit Product Comparison Guide
General description
Nuts and ferrules included.
Reducing unions of Swagelok® type are mostly used as metal column terminators. All original reducing unions have a bore in the centre of about 6mm in length and an internal diameter of 1.2mm. Moreover, reducing unions have a large bore at the non-reducing site. Modification is necessary in reducing union in order to minimize peak broadening, pressure resistance and to ease the installation process.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service