Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21008-U

Supelco

Stainless Steel Screen

Synonym(s):

Large PUF Cartridges

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
$21.10

$21.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 EA
$21.10

About This Item

UNSPSC Code:
12190000
NACRES:
NB.24

$21.10


Please contact Customer Service for Availability

Request a Bulk Order

product line

ORBO

packaging

pkg of 2 ea

manufacturer/tradename

Supelco ORBO 2000

technique(s)

active air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

Stainless Steel Screens are useful for ASTM D6209, EPA IP-7, and EPA TO-13 (PAHs); EPA TO-4A (Pesticides/PCBs); EPA TO-9A (Dioxins).

Legal Information

ORBO is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service