Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20640-U

Supelco

SNOOP® Liquid Leak Detector

1gal. (3.8 L) bottle

Synonym(s):

Leak Detection Fluid

Sign Into View Organizational & Contract Pricing

Select a Size

3.8 L
$377.00

$377.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
3.8 L
$377.00

About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

$377.00


Please contact Customer Service for Availability

Request a Bulk Order

General description

Liquid leak detectors are suitable to detect leaks. It is easy to use and inexpensive. When squirted on fitting, a leak becomes visible by the continuous formation of bubbles in the liquid. It is recommended to be used for leakage detection of fittings which have temperature below 100°C.[1]

Other Notes

For use with volumetric (bubble) flowmeters. Also useful for checking gas delivery system plumbing for leaks. Not recommend for use upstream of capillary GC systems (an electronic leak detector is preferred).

Legal Information

SNOOP is a registered trademark of Swagelok Company

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dean Rood
The Troubleshooting and Maintenance Guide for Gas Chromatographers, 201-201 (2007)

Questions

  1. I need the MSDS for the following: 20640-U Supelco SNOOP® Liquid Leak Detector

    1 answer
    1. The SDS for this product is published on the website. A link to this document is in the upper left of the product page, just below the thumbnail image.

      Please see a link to this document below:
      https://www.sigmaaldrich.com/sds/supelco/20640-u

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service