Key Documents
20396
Supelco
Column Tag
engraved
, two straps, pkg of 50 ea
Select a Size
50 EA
$148.00
$148.00
Available to ship onMarch 31, 2025Details
Request a Bulk Order
Select a Size
Change View
50 EA
$148.00
About This Item
UNSPSC Code:
41121800
$148.00
Available to ship onMarch 31, 2025Details
Request a Bulk Order
packaging
pkg of 50 ea
technique(s)
gas chromatography (GC): suitable
General description
These aluminum tags are easily marked with a Burgess Vibrograver or other scribing tool, for a permanent record of important chromatographic data.
Choose from one of the most recent versions:
Certificates of Analysis (COA)