Skip to Content
MilliporeSigma
All Photos(2)

Documents

20366-U

Supelco

ORBO 91T Carbotrap B / Carbosieve SIII Sorbent Tube (3-bed)

O.D. × L 7 mm × 150 mm, pkg of 25 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000

material

W,W,W separators

composition

Bed A, 100 mg
Bed B, 200 mg (Bed C 100mg)
Bed C, 100 mg

packaging

pkg of 25 ea

manufacturer/tradename

ORBO 91T

O.D. × L

7 mm × 150 mm

matrix

Carbosieve S-III Carbon Molecular Sieve (CMS) (Bed B & Bed C)
Carbotrap® B Graphitized Carbon Black (GCB) (Bed A)

application(s)

air monitoring

compatibility

for sampling Volatile Organic Compounds (VOCs) C2-C12 (Japanese methodology)

Legal Information

Carbotrap is a registered trademark of Merck KGaA, Darmstadt, Germany
ORBO is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service