Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20078-U

Supelco

Modified BTEXTRAP L, Tekmar® 3000

Synonym(s):

Modified BTEX Trap L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NB.21

application(s)

environmental

compatibility

for use with Tekmar® 3000, 3100 (Straight, 12 in./30.5 cm, Valco fitting

Looking for similar products? Visit Product Comparison Guide

Legal Information

BTEXTRAP is a trademark of Sigma-Aldrich Co. LLC
Tekmar is a registered trademark of Tekmar Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service