Skip to Content
MilliporeSigma
All Photos(1)

Documents

20044

Supelco

ORBO 353 HBr on Petroleum Charcoal (20/40), 100/50 mg

W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900

material

W,W,W separators

Agency

NIOSH 1614
OSHA 50,97

product line

ORBO

composition

Bed A, 100 mg
Bed B, 50 mg

packaging

pkg of 25 ea

manufacturer/tradename

ORBO 353

technique(s)

active air sampling: suitable

O.D. × L

6 mm × 75 mm

matrix

HBr on petroleum charcoal

particle size

20-40 mesh

application(s)

air monitoring
environmental
industrial hygiene

General description

  • ORBO 353 HBr on Petroleum Charcoal (20/40), 100/50 mg solvent desorption tube is designed for gas and vapor sampling.
  • It contains two beds of the same selective adsorbent separated by glass wool or foam.
  • Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.

Legal Information

ORBO is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service