Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1899-1AMP

Supelco

F.A.M.E. Mix GLC-100

certified reference material, ampule of 100 mg (Neat)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 100 mg (Neat)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

multi-component solution

functional group

carboxylic acid
ester

shipped in

dry ice

storage temp.

-10 to -25°C

General description

This quantitative mix is useful for determining relative retention times and approximating response factors.

Application

GLC-10 FAME mix was used as analytical standard for GC-FID analysis to construct calibration curve in determining the FAME component in biodiesel fuel.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Each lot is prepared by weight percent.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Methyl arachidate 20 wt. %
    Methyl behenate 20 wt. %
    Methyl heneicosanoate 20 wt. %
    Methyl nonadecanoate
    Methyl stearate 20 wt. %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanism and kinetics of thermal decomposition of biodiesel fuel.
Lin, Ronghong, Yiying Zhu, and Lawrence L. Tavlarides.
Fuel: The Science and Technology of Fuel and Energy, 106, 593-604 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service