Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13993-U

Supelco

Packed GC Column Set

suitable for 2261 per GPA (3-column set)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
eCl@ss:
32110502
NACRES:
SB.54

Agency

suitable for GPA 2261 (3-column set)

technique(s)

gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

column type

packed GC

Looking for similar products? Visit Product Comparison Guide

General description

This column set is specifically designed for a specific column switching application. Column manufacturing procedurs have been optimized resulting in column sets that provide highly reproducible analyses.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Introduction to Analytical Gas Chromatography.
Scott RPW.
Science, 101-104 (1997)
Capillary and packed-column gas chromatography of gasoline hydrocarbons and EDB.
Bruell CJ and Hoag GE.
Science, 5-7 (1984)

Protocols

Packed columns for natural gas BTU determination as per GPA and ASTM methods, ensuring accurate measurements in gas processing.

Chromatograms

suitable for GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service