Skip to Content
MilliporeSigma
All Photos(1)

Documents

13014-U

Supelco

Metal Packed GC Column (fits Agilent, Configuration "A")

phase 20% Sebaconitrile, matrix 80/100 Chromosorb P AW support, L × O.D. × I.D. 2.0 ft (0.61 m) × 1/8 in. × 2.1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
NACRES:
SB.54

material

stainless steel column

technique(s)

gas chromatography (GC): suitable

L × O.D. × I.D.

2.0 ft (0.61 m) × 1/8 in. × 2.1 mm

matrix

80/100 Chromosorb P AW support

matrix active group

20% Sebaconitrile phase

column type

packed GC

compatibility

for use with Agilent Configuration A

Looking for similar products? Visit Product Comparison Guide

General description

The carrier gas percolates through the porous bed in packed columns. In a packed column, separation is achieved by moving the solutes apart. Packed columns are made up of mainly two materials; stainless steel and Pyrex glass. Metal packed columns are more suitable for long and high carrier gas flow rates to tolerate the elevated pressures. Chromosorb P is a diatomite support to any stationary phase. Chromosorb P (AW) gives the best separation of volatile fatty acids consistent with the lowest temperatures.
An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column fits Agilent 5880, 5890, 6890, 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Lungs, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Remesy et al.
The Biochemical journal, 141(1), 85-91 (1974-07-01)
1. A new rapid micro-method for measuring plasma volatile fatty acids is described. The volatile fatty acids are extracted from plasma with ethanol in the presence of a known quantity of internal standard (sodium isobutyrate). After evaporation of the ethanolic
Raymond P. W. Scott
Introduction to Analytical Gas Chromatography (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service