Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11993

Supelco

Coated GC Packing

phase 5% Carbowax 20M, matrix 40/60 Chromosorb T support, bottle of 50 g

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$2,590.00

$2,590.00


Please contact Customer Service for Availability


Select a Size

Change View
50 G
$2,590.00

About This Item

UNSPSC Code:
23151817

$2,590.00


Please contact Customer Service for Availability

packaging

bottle of 50 g

parameter

60-225 °C temp. range

technique(s)

gas chromatography (GC): suitable

matrix

40/60 Chromosorb T support

matrix active group

5% Carbowax 20M phase

column type

packed GC

compatibility

for analyte group general

General description

Ready-to-use coated packing for those wanting to pack their own columns.

Application

general use

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Columns for Gas Chromatography: Performance and Selection.
Grob RL and Barry EF.
Science, 22-23 (2007)
Analytical Measurements in Aquatic Environments.
Namiesnik J and Szefer P.
Science, 73-73 (2009)

Questions

1–4 of 4 Questions  
  1. How can I purchase a column that is packed with this material?

    1 answer
    1. Most packed column are custom made and require that you obtain a quotation. Please contact our Customer & Sales Services group at phone number (800)-247-6628, fax number (800)-447-3044 or email [email protected] for additional information.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. Is this product a packed column?

    1 answer
    1. This is packing material only; it is not a column. Some endusers will purchase this bottle of packing and prepare their own packed columns.

      Helpful?

  4. What is the maximum temperature for this packing?

    1 answer
    1. The maximum temperature for this packing is 225 °C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service