Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11045

Supelco

GC Stationary Phase

phase Diethylene glycol succinate (DEGS), bottle of 25 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

Agency

meets requirements for USP G4

packaging

bottle of 25 g

parameter

temp. range at 20-200 °C
20-200 °C temp. range

technique(s)

gas chromatography (GC): suitable

matrix active group

Diethylene glycol succinate (DEGS) phase

solubility

acetone: soluble

column type

packed GC

Looking for similar products? Visit Product Comparison Guide

General description

In gas chromatography (GC) solutes are sorbed and desorbed only in the stationary phase. There is a large variety of stationary phases and the choice of it depends on its ability to interact with the sample. Diethylene glycol succinate stationary phase is a non-volatile liquid phase.
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Application

Diethylene glycol succinate was used as a stationary phase for the separation of glyceryl ethers from the liver oils of the grey dogfish, the Greenland shark, and the ratfish.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Basics of Analytical Chemistry and Chemical Equilibria.
Tissue B M.
Science, 10-10 (2013)
Gas chromatography.
Singh R.
Science, 60-61 (2002)
Chromatography: Concepts and Contrasts.
Miller J M.
Science, 102-103 (2005)
The glyceryl ethers in the liver oils of elasmobranch fish.
Bo H and Larsson S.
Journal of Lipid Research, 3 (1), 31-38 (1962)
Kai Wang et al.
Biochemical pharmacology, 155, 494-509 (2018-08-03)
The present study aimed to evaluate the anti-colitis effect and underlying mechanisms of cardamonin, a natural flavone isolated from Alpinia katsumadai Hayata. The results showed that oral cardamonin significantly inhibited dextran sulfate sodium (DSS)- and 2,4,6-trinitrobenzene sulfonic acid (TNBS)-induced colitis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service