Skip to Content
MilliporeSigma
All Photos(17)

Documents

ZRB939

Sigma-Aldrich

Anti-Pancreatic Polypeptide Antibody, clone 3H5, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Obinepitide, PP, Pancreatic Prohormone

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3H5, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 10.44 kDa

species reactivity

human

species reactivity (predicted by homology)

bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable using 1:100
immunofluorescence: 1:100-1:400
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:400
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PPY;PNP(5539)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 3H5 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human Pancreatic Polypeptide.

Immunogen

KLH-conjugated linear peptide corresponding to full-length human Pancreatic Polypeptide (without the signal and propeptides).

Application

Anti-Pancreatic Polypeptide, clone 3H5 ZooMAb, Cat. No. ZRB939, is a recombinant Rabbit Monoclonal Antibody that detects pancreatic polypeptide and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, and Western Blotting.
Immunofluorescence Analysis: A 1:400 dilution from a representative lot detected Pancreatic Polypeptide in human pancreas tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:400 dilution from a representative lot detected Pancreatic Polypeptide in human pancreas tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Pancreatic Polypeptide in TGP52 cell line.

Target description

Pancreatic polypeptide (UniProt: P01298; also known as PP, Obinepitide, Pancreatic prohormone) is encoded by the PPY (also known as PNP) gene (Gene ID: 5539) in human. Pancreatic polypeptide is a member of the neuropeptide Y family and is secreted protein that is produced by PP cells in the pancreatic islets of Langerhans. The pancreatic prohormone is synthesized with a signal peptide (aa 1-29) and a propeptide (aa 89-95) that are subsequently cleaved off. The remaining part is cleaved off into pancreatic hormone (aa 30-65) and pancreatic icosapeptide (69-88). Pancreatic hormone acts as a regulator of pancreatic and gastrointestinal functions. Two isoforms of pancreatic polypeptide have been described that are generated by alternative splicing. Pancreatic polypeptide release is functionally associated with increased satiety, delayed gastric emptying, inhibition of gallbladder motility, and regulation of energy expenditure. Its deficiency is observed in cases of pancreatic resection and chronic pancreatitis and is associated with significant reduction in PP cell mass and abnormally high hepatic glucose production even in the presence of endogenous insulin. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Maxwell, J et al. (2014). Pancreas 43(4); 651-656).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

120 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service