Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

ZRB3155

Sigma-Aldrich

Anti-TRPV4 Antibody, clone 1G10-L2 ZooMAb® Rabbit Monoclonal

newenhanced validation

recombinant, expressed in HEK 293 cells

Synonym(s):

OTRPC4, Osm-9-like TRP channel 4, TRP12, Transient receptor potential cation channel subfamily V member 4, Transient receptor potential protein 12, TrpV4, VRL-2, Vanilloid receptor-like channel 2, Vanilloid receptor-like protein 2, Vanilloid receptor-related osmotically-activated channel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G10-L2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 98.28 kDa
observed mol wt ~110 kDa

purified by

using Protein A

species reactivity

human, mouse

species reactivity (predicted by homology)

rat, canine

packaging

antibody small pack of 25

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... TRPV4(59341)

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1G10-L2 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects TRPV4. It targets an epitope within 17 amino acids from an extracellular domain in the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from an extracellular domain in the C-terminal half of human TRPV4.

Application

Quality Control Testing

Evaluated by Western Blotting in A-375 cell lysate.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected TRPV4 in A-375 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected TRPV4 in lysate from mouse testis tissue.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected TRPV4 in human kidney tissue sections, and a 1:100 dilution of the same lot detected TRPV4 in human heart tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound TRPV4 peptide with at least ten thousand-fold (10,000X) higher than with non-specific control peptide.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TRPV4 in A-375 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Transient receptor potential cation channel subfamily V member 4 (UniProt: Q9HBA0; also known as TrpV4, Osm-9-like TRP channel 4, OTRPC4, Transient receptor potential protein 12, TRP12, Vanilloid receptor-like channel 2, Vanilloid receptor-like protein 2, VRL-2, Vanilloid receptor-related osmotically-activated channel, VR-OAC) is encoded by the TRPV4 (also known as VRL2, VROAC) gene (Gene ID: 59341) in human. It belongs to the OSM9-like transient receptor potential channel (OTRPC) subfamily within the transient receptor potential (TRP)3 superfamily of channels. TRPV4 is a multi-pass membrane protein characterized by six transmembrane domains (S1-S6) and a pore-forming loop between S5 and S6. It also has three ankyrin repeat domains (ANK 1-3) which are crucial for protein-protein interactions and channel regulation, and mediates interaction with Ca2+-calmodulin and ATP binding. TRPV4 is widely expressed in various tissues including endothelial cells, sensory neurons, cartilage, bone and epithelial cells of the kidney, lung, and skin. It is also known to be found in the synoviocytes from patients with (RA). TRPV4 is primarily localized to the plasma membrane, and can also be found in intracellular compartments such as the endoplasmic reticulum, particularly in cells where it plays a role in calcium signaling and homeostasis. Six isoforms of TRPV4 have been described that are produced by alternative splicing. Mutations or dysregulation of TRPV4 are linked to several diseases like skeletal dysplasia, pulmonary edema, pain and Inflammation and cardiovascular diseases. ANK domain mutations are found in patients suffering from spinal muscular atrophies (SMA, also known as hereditary motor neuropathies) and hereditary motor and sensory neuropathies (HMSN). This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service