Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZRB2714

Sigma-Aldrich

Anti-TrkB Antibody, clone 3E18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

BDNF/NT-3 growth factors receptor;EC:2.7.10.1;GP145-TrkB;Trk-B;Neurotrophic tyrosine kinase receptor type 2;TrkB tyrosine kinase;Tropomyosin-related kinase B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3E18, recombinant monoclonal

description

3E18 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 93.45 kDa

purified by

using Protein A

species reactivity

human, mouse

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable

isotype

IgG

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NTRK2(4915)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3E18 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects TrKB. It targets an epitope within 18 amino acids from the extracellular domain within the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the extracellular domain within the N-terminal half.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in Neuro 2A cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected TrkB in Neuro 2A cells.

Tested applications

Affinity Binding Assay: A representative lot of this antibody bound TrkB peptide a KD of 1.6 x 10-6 in an affinity binding assay.

Enzyme Immunoassay Analysis (ELISA): Serial of dilutions of this antibody detected recombinant human TrkB.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Immunocytochemistry in Neuro 2A cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected TrkB in Neuro 2A cells.

Target description

BDNF/NT-3 growth factors receptor (UniProt: Q16620; also known as EC:2.7.10.1, GP145-TrkB, Trk-B, Neurotrophic tyrosine kinase receptor type 2, TrkB tyrosine kinase, Tropomyosin-related kinase B) is encoded by the NTRK2 (also known as TRKB) gene (Gene ID: 4915) in human. TrkB is a single-pass type I membrane glycoprotein that is internalized to endosomes upon ligand-binding. It is synthesized with a signal peptide (aa 1-31) that is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 32-430), a transmembrane domain (aa 431-454), and a cytoplasmic domain (aa 455-822). TrkB is a receptor tyrosine kinase that is involved in the development and maturation of the central and the peripheral nervous systems through regulation of neuron survival, proliferation, migration, differentiation, and synapse formation and plasticity. In the central nervous system, its expression is observed in the cerebral cortex, hippocampus, thalamus, choroid plexus, granular layer of the cerebellum, brain stem, and spinal cord. In the peripheral nervous system, it is expressed in many cranial ganglia, the ophthalmic nerve, the vestibula, and multiple facial structures. Its expression is highly regulated and determines the sensitivity of neurons to BDNF. Upon ligand-binding, TrkB undergoes homodimerization, autophosphorylation, and activation. It can then recruit, phosphorylate and/or activate several downstream effectors including SHC1, FRS2, SH2B1, SH2B2 and PLCG1 that regulate distinct overlapping signaling cascades. TrkB is also reported to play a role in neutrophin-dependent calcium signaling in glial cells and mediate communication between neurons and glia. Following binding of BDNF to TrkB receptor, Tyrosine 816 is phosphorylated, which allows the binding of phospholipase C and activation of signaling cascade. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Andreska, T., et al. (2020). Cell Tissue Res. 382(1); 5-14; Klein, R., et al. (1991). Cell. 66(2); 395-403).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service