Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

ZRB2595

Sigma-Aldrich

Anti-S100A12 Antibody, clone 1L6 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CAAF1, CAGC, Extracellular newly identified RAGE-binding protein, CGRP, Calcium-binding protein in amniotic fluid 1, Calgranulin-C, EN-RAGE, MRP-6, Migration inhibitory factor-related protein 6, Neutrophil S100 protein, S100 calcium-binding protein A12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1L6
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 10.58 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... S100A12(6283)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1L6 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects S100A12 protein. It targets an epitope within 22 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the internal region of human S100A12.

Application

Quality Control Testing

Evaluated by Western Blotting with full-length recombinant Human S100A12.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected full-length recombinant Human S100A12.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution of this antibody detected S100A12 in Human bone marrow tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound S100A12 peptide with a KD of 1.8 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected S100A12 in HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user..

Target description

Protein S100-A12 (UniProt: P80511; also known as CGRP, Calcium-binding protein in amniotic fluid 1, CAAF1, Calgranulin-C, CAGC, Extracellular newly identified RAGE-binding protein, EN-RAGE, Migration inhibitory factor-related protein 6, MRP-6, Neutrophil S100 protein, S100 calcium-binding protein A12) is encoded by the S100A12 gene (Gene ID: 6283) in human. S100A12 is a member of the S100 family of calcium-binding proteins that exists as a homodimer but can form tetramers or hexamers in the presence of calcium, zinc, and copper ions. It is predominantly expressed by neutrophils, monocytes, and activated macrophages. Its expression is also observed in eosinophils and macrophages in asthmatic airways in regions where mast cells accumulate. Its best-known target is RAGE (receptor for advanced glycation end products). It is predominantly localized in the cytoplasm, however, upon elevation of the intracellular calcium level, it translocates from the cytoplasm to the cytoskeleton and the cell membrane. It contains two EF-hand domains (aa 13-48 and 49-84) and a hinge region (aa 38-53). Each EF-domain can bind to one calcium ion that changes its conformation resulting in a concave hydrophobic surface in the C-terminal region. This forms a target recognition site thus enabling selective interaction with a host of specific protein or peptide targets. S100A12 plays a prominent role in the regulation of inflammatory processes and immune response. Its pro-inflammatory activity involves recruitment of leukocytes, promotion of cytokine and chemokine production, and regulation of leukocyte adhesion and migration. It acts as an alarmin, or a danger associated molecular pattern (DAMP) molecule and stimulates innate immune cells via binding to receptor for advanced glycation end products (AGER). Binding to AGER activates the MAP-kinase and NF-kB signaling pathways leading to production of pro-inflammatory cytokines and up-regulation of cell adhesion molecules ICAM1 and VCAM1. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Meijer, B., et al. (2012). Int. J. Inflam. 2012: 907078; Moroz, MV., et al. (2009). BMC Biochem. 10; 11; Yang, Z., et al. (2007). J. Allergy Clin. Immunol. 119(1); 106-114).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service