Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

ZRB2566

Sigma-Aldrich

Anti-HOPX Antibody, clone 2L10 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Homeodomain-only protein, Lung cancer-associated Y protein, Not expressed in choriocarcinoma protein 1, Odd homeobox protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2L10, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 8.26 kDa
observed mol wt ~11 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... HOPX(84525)

Related Categories

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2L10 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Homeodomain-only protein (HOPX).

Immunogen

His-tagged, full-length, recombinant human Homeodomain-only protein (HOPX).

Application

Quality Control Testing

Evaluated by Western Blotting in Human Placenta tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HOPX in Human placenta tissue lysate.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected HOPX in Human placenta tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound His-tagged, full-length, recombinant human HOPX with a KD of 5.6 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Homeodomain-only protein (UniProt: Q9BPY8; also known as HOPX, Lung cancer-associated Y protein, Not expressed in choriocarcinoma protein 1, Odd homeobox protein 1) is encoded by the HOPX (also known as HOD, HOP, LAGY, NECC1, OB1) gene (Gene ID: 84525) in human. HOPX is an atypical homeodomain protein that lacks the capacity to bind DNA and instead functions by interacting with transcriptional regulators. It plays crucial roles in organogenesis and its expression is observed in both embryonic and adult stem cells. It is reported to be important for anterior-posterior patterning during organ development and its functional impairment is associated with a variety of diseases, including cancer. It serves as a tumor suppressor that regulates cellular proliferation and metastasis. Down-regulation of HOPX is reported in lung cancer, choriocarcinoma, head and neck squamous cell carcinoma, and oral squamous cell carcinoma. It interacts with serum response factor (SRF) and modulates the expression of SRF-dependent cardiac-specific genes and cardiac development. It prevents SRF-dependent transcription either by inhibiting SRF binding to DNA or by recruiting histone deacetylases that prevent transcription by SRF. Four isoforms of HOPX have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Gokulan, RC., et al. (2022). Cancers (Basel). 14(11); 2764; Ota, C., et al. (2018). Sci. Rep. 8; Article 12983).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service