Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ZRB2553

Sigma-Aldrich

Anti-FIS1 Antibody, clone 1C7 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CGI-135, FIS1 homolog (hFis1), Mitochondrial fission 1 protein, TPR repeat protein 11, TTC11, Tetratricopeptide repeat protein 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C7, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 16.9 kDa
observed mol wt ~17 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

monkey, bovine, rat, horse

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

N-terminal cytoplasmic domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... FIS1(51024)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C7 is a ZooMAb® rabbit recombinant monoclonal antibody that detects FIS1. It targets an epitope within 24 amino acids from the N-terminal cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal, cytoplasmic domain of human FIS1.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting: A 1:10,000 dilution of this antibody detected FIS1 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: 1:1,000-10,000 dilutions from a representative lot detected FIS1 in lysates from Raji cells and mouse liver tissue.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected FIS1 in human kidney tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected FIS1 in HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Mitochondrial fission 1 protein (UniProt: Q9Y3D6; also known as FIS1 homolog (hFis1), Tetratricopeptide repeat protein 11 (TPR repeat protein 11)) is encoded by the FIS1 (also known as TTC11, CGI-135) gene (Gene ID: 51024) in human. FIS1 is a small, integral mitochondrial outer membrane protein characterized by a tetratricopeptide repeat (TPR) domain which facilitates protein-protein interactions. The cytosolic domain interacts with other proteins involved in mitochondrial fission. It acts by recruiting cytosolic factors such as dynamin-related protein 1 (DRP1), facilitating the assembly of the fission machinery that constricts and divides mitochondria. This process is crucial for mitochondrial distribution and health, affecting cellular energy production and apoptosis. Dysregulation of FIS1 expression or activity is associated with neurodegenerative diseases such as Alzheimer′s and Parkinson′s, where impaired mitochondrial dynamics contribute to cellular dysfunction and death. Additionally, FIS1 has been linked to metabolic disorders, as proper mitochondrial function is crucial for energy balance and metabolism. In cancer, altered mitochondrial fission can affect apoptosis and cellular proliferation, implicating FIS1 in tumorigenesis and cancer progression. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service