Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

ZRB2532

Sigma-Aldrich

Anti-S100-A9/MRP14 Antibody, clone 1M6 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Calgranulin-B, Calprotectin L1H subunit, Leukocyte L1 complex heavy chain, MRP-14, Migration inhibitory factor-related protein 14, S100 calcium-binding protein A9, p14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1M6, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 13.24 kDa
observed mol wt ~13 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... S100A9(6280)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1M6 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects S100-A9. It targets an epitope within 24 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal half of human S100-A9/MRP14.

Application

Quality Control Testing

Evaluated by Western Blotting in human tonsil tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected S100-A9/MRP14 in human tonsil tissue lysate.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound S100-A9/MRP14 peptide with at least one thousand-fold (1,000X) higher than with non-specific control peptide.

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected S100-A9/MRP14 in human tonsil tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected S100-A9/MRP14 in human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Protein S100-A9 (UniProt P06702; also known as Calgranulin-B, Calprotectin L1H subunit, Leukocyte L1 complex heavy chain, Migration inhibitory factor-related protein 14, MRP-14, p14, S100 calcium-binding protein A9, S100A9) is encoded by the S100A9 (also known as CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MRP14) gene (Gene ID 6280) in human. S100A9 is one of the 21 members of the S100 family of proteins. S100A8 (calgranulin-A), S100A9 (calgranulin-B) and the heterodimer S100A8/S100A9 (calprotectin) exert their immunological and inflammatory action via binding to TLR-4, RAGE and some other putative receptors. Predominantly, the protein is localized in the cytoplasm and with increased intracellular calcium levels it is translocated from the cytoplasm to the cytoskeleton and the cell membrane. Calprotectin (S100A8/9) is predominantly expressed in myeloid cells with exception in inflammatory conditions where the expression is restricted to a specific stage of myeloid differentiation and both proteins are expressed in circulating neutrophils and monocytes but are absent in normal tissue macrophages and lymphocytes. S100A9 is also reported to induce neutrophil chemotaxis and adhesion and can increase the bactericidal activity of neutrophils by promoting phagocytosis via activation of SYK, PI3K/AKT, and ERK1/2 pathways and can induce degranulation of neutrophils by a MAPK-dependent mechanism. Thus, S100A9 is a critical protein in both normal immune function and pathophysiology, serving as a potential therapeutic target for treating inflammatory conditions. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Atallah, M., et al. (2012). PloS One. 7(2); e29333; Nakatani, Y., et al. (2005). Mediators Inflamm. 2005(5); 280-292; Ryckman, C., et al. (2003). J. Immunol. 170(6); 3233-3242).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service