Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

ZRB2513

Sigma-Aldrich

Anti-MYH6 Antibody, clone 1L5 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Myosin heavy chain 6, Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha), Myosin-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1L5, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 223.74 kDa
observed mol wt ~224 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... MYH6(4624)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1L5 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects MYH6. It targets an epitope within 17 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal half of human MYH6.

Application

Quality Control Testing

Evaluated by Western Blotting in human heart tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MYH6 in human heart tissue lysate.

Tested Applications

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected MYH6 in human heart tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected MYH6 in human heart tissue sections and human skeletal muscle tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound MYH6 peptide with at least one hundred thousand-fold (100,000X) higher than with non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Myosin-6 (UniProt: P13533; also known as Myosin heavy chain 6, Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha)) is encoded by the MYH6 (also known as MYHCA) gene (Gene ID:4624) in human. MYH6 (myosin, heavy chain 6) gene encodes the heavy chain subunit of the cardiac muscle myosin, which is a hexamer made up of two heavy chain subunits, two light chain subunits, and two regulatory subunits. It is involved in the contraction of muscles. MYH6 is predominantly expressed in the heart, particularly in the atrial muscle. It is one of the major isoforms of cardiac myosin heavy chain, with its expression being more significant in the atria compared to the ventricles. MYH6 is localized in the sarcomere, the fundamental unit of muscle contraction, within the cardiac muscle cells. Specifically, it is a component of the thick filaments in the A-band of the sarcomere, where it interacts with actin filaments to facilitate muscle contraction. Defects in this gene cause atrial septal defect 3 (ASD3) characterized by incomplete closure of the wall between the atria resulting in blood flow from the left to the right atria. Its expression is also altered during thyroid hormone depletion and hemodynamic overloading. Mutations in MYH6 can disrupt the normal function of the protein, impairing cardiac muscle contraction and leading to several cardiac diseases. Mutations in this gene are associated with familial hypertrophic cardiomyopathy, dilated cardiomyopathy, and heart rhythm disorders due to malfunction of the sinus node, leading to irregular heartbeats. Genetic screening and molecular studies are crucial for identifying these mutations and understanding their impact on cardiac function. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service