Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ZRB2510

Sigma-Aldrich

Anti-GPx4 Antibody, clone 1G5 ZooMAb® Rabbit Monoclonal

newenhanced validation

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:1.11.1.12, EC:1.11.1.9, GPx-4, GSHPx-4, Glutathione Peroxidase 4, PHGPx, Phospholipid hydroperoxide glutathione peroxidase GPX4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G5, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 22.18 kDa
observed mol wt ~18 kDa

purified by

using Protein A

species reactivity

human, mouse, rat

species reactivity (predicted by homology)

monkey, chimpanzee

packaging

antibody small pack of 25

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1G5 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects GPx4. It targets an epitope within 22 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the C-terminal region of human GPX4.

Application

Quality Control Testing

Evaluated by Western Blotting in U-937 cell lysate.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected GPx4 in U-937 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000-10,000 dilution from a representative lot detected GPx4 in lysate from mouse liver and rat testis tissue.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected GPx4 in human testis tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound GPx4 peptide with at least ten thousand-fold (10,000X) higher than with non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Phospholipid hydroperoxide glutathione peroxidase, cytosolic (UniProt: P36969; also known as PHGPx, Glutathione peroxidase 4, GPx-4, GSHPx-4) is encoded by the GPX4 gene (Gene ID: 2879) in human. The protein encoded by this gene belongs to the glutathione peroxidase family, members of which catalyze the reduction of hydrogen peroxide, organic hydroperoxides and lipid hydroperoxides, and thereby protect cells against oxidative damage. This isozyme is also a selenoprotein, containing the rare amino acid selenocysteine (Sec) at its active site. This selenocysteine is crucial for the enzyme′s catalytic activity. Sec is encoded by the UGA codon, which normally signals translation termination. Three functionally distinct isozymes of GPx4 have been described and are known as mGPx4 (mitochondrial), cGPx4 (cytosolic), and nGPx4 (nuclear). They originate from a single GPX4 gene. The cytosolic isoform is ubiquitously expressed at moderate levels in most mammalian cells. On the other hand, mitochondrial and nuclear isoforms are overexpressed in spermatoid cells. GPx4 is a considered to be a ′moonlighting′ protein because of its ability to serve dual functions as a peroxidase, that has been implicated in anti-oxidative defense, gene expression regulation, and programmed cell death, and as a structural protein in spermatogenesis. Mutations or deficiencies in GPX4 are associated with several diseases. One of the most severe conditions linked to GPX4 deficiency is Sedaghatian-type spondylometaphyseal dysplasia (SMDS), a rare genetic disorder characterized by skeletal abnormalities, neurological impairment, and early infant death. Homozygous GPx4-deficient mice are reported to die utero before E7.5. Similarly, homozygous mice expressing catalytically inactive GPx4 variants (Sec46Ala/Ser mutant) do not survive beyond the 7th day of embryogenesis. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Borchert, A., et al (2010). Immunol. Lett. 133 (2); 85-93).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service