Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

ZRB2396

Sigma-Aldrich

Anti-MST1/STK4 Antibody, clone 1L17-L2 ZooMAb® Rabbit Monoclonal

enhanced validation

recombinant, expressed in HEK 293 cells

Synonym(s):

EC: 2.7.11.1, Mammalian STE20-like protein kinase 1, STE20-like kinase MST1, Serine/threonine-protein kinase 4, Serine/threonine-protein kinase Krs-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1L17-L2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... STK4(6789)

Related Categories

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1L17-L2 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Serine/threonine-protein kinase 4 (MST1/STK4). It targets an epitope within 16 amino acids from the N-terminal region. Detects the intact and cleaved forms.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal region of human Serine/threonine-protein kinase 4 (MST1/STK4).

Application

Quality Control Testing

Evaluated by Western Blotting in Ramos cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected MST1/STK4 in Ramos cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected MST1/STK4 in Caco2 cell lysate.

Affinity Binding Assay: A representative lot of this antibody bound MST1/STK4 peptide with a KD of 1.8x 10-6 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected MST1/STK4 in Human tonsil tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MST1/STK4 in Ramos cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Serine/threonine-protein kinase 4 (UniProt: Q13043; also known as EC: 2.7.11.1, Mammalian STE20-like protein kinase 1, MST-1, STE20-like kinase MST1, Serine/threonine-protein kinase Krs-2, STK4) is encoded by the STK4 (also known as KRS2, MST1) gene (Gene ID: 6789) in human. STK4 is a homodimeric, stress-activated, pro-apoptotic kinase that enters the nucleus following its caspase cleavage and induces chromatin condensation followed by internucleosomal DNA fragmentation. It is ubiquitously expressed with higher expression observed in prostate cancer and its levels increase with the progression to malignant state. Its protein kinase domain is localized in residues 30-281. It is activated by caspase-cleavage between amino acids 326-327 and 349-350. The caspase-cleaved form cycles between the nucleus and cytoplasm. Its full activation requires homodimerization and autophosphorylation at threonine 183. Its activity is inhibited by the C-terminal non-catalytic region. STK4 is a KEY® component of the Hippo signaling pathway, which plays a pivotal role in organ size control and acts as a tumor suppressor by restricting cell proliferation and promoting apoptosis. During apoptosis, it phosphorylates serine 14 of histone H2B and phosphorylates FOXO3 upon oxidative stress that results in its NuCLEAR translocation and initiation of cell death. Its phosphorylation at threonine 387 by AKT1, LEADS to inhibition of its kinase activity, NuCLEAR translocation and autophosphorylation at threonine 183. It also reduces its cleavage by caspases and its ability to phosphorylate FOXO3. Two isoforms of STK4 have been described that are produced by alternative splicing. Mutations in STK4 gene have been linked to T cell immunodeficiency resulting from progressive loss of naive T-cells. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: (Ref.: Guennoun, A., et al. (2021). J. Clin. Immunol. 41(8); 1839-1852; Praskova, M., et al. (2004). Biochem. J. 381(Pt2); 453-462; Lee, KK., et al. (2001). J. Biol Chem. 276(22); 19276-19285).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
KEY is a registered trademark of Hanna Instruments
LEADS is a trademark of Compugen
NuCLEAR is a trademark of Sigma-Aldrich Co. LLC
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service