Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

ZRB2351

Sigma-Aldrich

Anti-USP11 Antibody, clone 1E15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Deubiquitinating enzyme 11, EC:3.4.19.12, Ubiquitin carboxyl-terminal hydrolase 11, Ubiquitin thioesterase 11, Ubiquitin-specific-processing protease 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E15, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... USP11(8237)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E15 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Ubiquitin carboxyl-terminal hydrolase 11 (USP11). It targets an epitope within 25 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the N-terminal half of human Ubiquitin carboxyl-terminal hydrolase 11 (USP11).

Application

Quality Control Testing

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected USP11 in HepG2 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected USP11 in HeLa cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected USP11 in Human cerebral cortex tissue sections .

Affinity Binding Assay: A representative lot of this antibody bound USP11 peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Ubiquitin carboxyl-terminal hydrolase 11 (UniProt: P51784; also known as EC:3.4.19.12, Deubiquitinating enzyme 11, Ubiquitin thioesterase 11, Ubiquitin-specific-processing protease 11) is encoded by the USP11 (also known as UHX1) gene (Gene ID: 8237) in human. USP11 is a monomeric protein of the peptidase C19 family that can remove conjugated ubiquitin from target proteins and polyubiquitin chains. It is mainly localized in the nucleus with only negligible part present in the cytoplasm. It inhibits the degradation of target proteins by the proteasome. It is involved in the regulation of multiple processes in cells, including cell cycle, DNA repair, tumor development, and signaling. It also acts as a chromatin regulator via its association with the Polycomb group (PcG) multiprotein PRC1-like complex; may ACT® by deubiquitinating components of the PRC1-like complex. It is reported to deubiquitinate and stabilize NRF2, thereby regulating cell proliferation and ferroptotic cell death. Depletion of USP11 is known to suppression of cell proliferation and induce of ferroptotic cell death due to ROS-mediated stress that can be largely abrogated by overexpression of NRF2. Its USP domain is localized in amino acids 309-930. USP11 cleaves preferentially lysine 6- and lysine 63-linked ubiquitin chains and displays lower activity with lysine 11- and lysine 33-linked ubiquitin chains, and extremely low activity with lysine 27-, 29-, and 48-linked ubiquitin chains. Dysregulation of USP11 is observed in several human tumors, including melanoma, glioma, colorectal cancer, and cervical cancer. It is reported to promote tumorigenesis by stabilizing cellular inhibitor of apoptosis protein 2 in colon cancer and melanoma, however, it can also suppress tumorigenesis by stabilizing PTEN homolog in prostate cancer. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Guo, T., et al. (2022). J. Oncol. 2022; Article ID 9963905; Meng, C., et al. (2021). Oncogene. 40(9); 1706-1720; Harper, S., et al. (2014). Biochemistry. 53(18); 2966-2978).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ACT is a registered trademark of Merck KGaA, Darmstadt, Germany
Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service