Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

ZRB2333

Sigma-Aldrich

Anti-LNPEP/IRAP Antibody, clone 1G23 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cystinyl aminopeptidase, EC:3.4.11.3, IRAP, Insulin-regulated membrane aminopeptidase, Insulin-responsive aminopeptidase, Leucyl-cystinyl aminopeptidase, OTase, Oxytocinase, P-LAP, Placental leucine aminopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

Clone, 1G23

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 117.35 kDa
observed mol wt ~170 kDa

purified by

using Protein A

species reactivity

mouse, rat, human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Cytoplasmic domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... LNPEP(4012)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1G23 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Leucyl-cystinyl aminopeptidase (LNPEP). It targets an epitope within 16 amino acids from the cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the cytoplasmic domain of human Leucyl-cystinyl aminopeptidase (LNPEP).

Application

Quality Control Testing

Evaluated by Western Blotting in Ramos cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LNPEP/IRAP in Ramos cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected LNPEP/IRAP in lysates from HeLa cells, C6 cells, and Mouse heart tissue..

Affinity Binding Assay:: A representative lot of this antibody bound LNPEP/IRAP peptide with a KD of 3.9 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected LNPEP/IRAP in HeLa cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected LNPEP/IRAP in Human cerebral cortex tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Leucyl-cystinyl aminopeptidase (UniProt: Q9UIQ6; also known as EC:3.4.11.3, Cystinyl aminopeptidase, Insulin-regulated membrane aminopeptidase, Insulin-responsive aminopeptidase, IRAP, Oxytocinase, OTase, Placental leucine aminopeptidase, P-LAP) is encoded by the LNPEP (also known as OTASE) gene (Gene ID: 4012) in human. LENPEP/IRAP is a zinc-dependent aminopeptidase that cleaves vasopressin, oxytocin, dynorphin A and other peptide hormones and is involved in the inactivation of neuronal peptides. It is a single-pass type II membrane protein with a cytoplasmic domain (aa 1-110), a transmembrane domain (aa 111-131), and a large extracellular domain (aa 132-1025). It is highly expressed in placenta, heart, kidney, and small intestine. Lower levels are detected in neuronal cells, skeletal muscle, spleen, liver, testes, and colon. In the brain tissue only a membrane-bound form is reported. It resides in intracellular vesicles together with GLUT4. and can then translocate to the cell surface in response to insulin and/or oxytocin. Cleavage of membrane-bound LENPEP/IRAP between residues 154-155 generates a pregnancy serum form that plays a role in maintaining homeostasis during pregnancy. Three isoforms of LENPEP/IRAP have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Matsumoto, H., et al. (2001). Eur. J. Biochem. 268(11); 3259-3266; Tsujimoto, M., et al. (1992). Arch. Biochem. Biophys. 292(2); 388-392).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service