Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

ZRB2264

Sigma-Aldrich

Anti-Interleukin-4 Antibody, clone 2C14 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

B-cell stimulatory factor 1 (BSF-1), Binetrakin, Lymphocyte stimulatory factor 1, Pitrakinra

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C14, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 17.49 kDa
observed mol wt ~16 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... IL4(3565)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2C14 is a ZooMAb® rabbit recombinant monoclonal antibody that detects Interleukin-4. It targets an epitope within 25 amino acids from the C-terminal part.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the C-terminal part of human Interleukin-4.

Application

Quality Control Testing

Evaluated by Western Blotting with human recombinant Interleukin-4.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected human recombinant Interleukin-4 .

Tested Applications

Affinity Binding Assay Analysis: The binding affinity of this antibody to Interleukin-4 peptide was at least one thousand-fold (1,000X) higher than that obtained with the non-specific control peptide.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Interleukin-4 in MOLT-4 cells with and without treatment with TPA (40 nM, 6 hrs) and Ionomycin (2 uM 6 hrs) and then treated with Brefeldin A (1ug/mL) for the last 5 .

Enzyme Immunoassay (ELISA): A representative lot of this antibody detected Interleukin-4 protein in ELISA application.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Interleukin-4 (UniProt: P05112; also known as B-cell stimulatory factor 1 (BSF-1), Binetrakin, Lymphocyte stimulatory factor 1, Pitrakinra) is encoded by the IL4 gene (Gene ID: 3565) in human. Interleukin-4 is a pleiotropic cytokine with a well-defined helical bundle structure. It plays a crucial role in regulating immune responses, particularly in promoting the differentiation of naive T cells into Th2 cells and stimulating B cell growth and antibody production. Interleukin-4 is primarily produced by activated T cells, basophils, and mast cells, and its expression can be induced by various stimuli, including antigens, T cell receptor signaling, and cytokines such as IL-2 and IL-4 itself. Upon secretion, Interleukin-4 binds to its receptor, initiating signaling cascades that activate the JAK-STAT pathway and modulate gene expression. Dysregulation of Interleukin-4 has been linked to allergic diseases, such as asthma and atopic dermatitis, as well as autoimmune disorders and certain cancers, highlighting its significance as a therapeutic target in these conditions. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service