Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB2109

Sigma-Aldrich

Anti-GRP75 Antibody, clone 2D20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Stress-70 protein mitochondrial;75 kDa glucose-regulated protein;GRP-75;Heat shock 70 kDa protein 9;Mortalin;MOT; Peptide-binding protein 74;PBP74

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2D20, recombinant monoclonal

description

2D20 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 73.68 kDa
observed mol wt ~70 kDa

purified by

using Protein A

species reactivity

rat, mouse, monkey, human

species reactivity (predicted by homology)

feline, canine, bovine, porcine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... HSPA9(3313)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2D20 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects GRP75. It targets an epitope within 20 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal half of human GRP75.

Application

Quality Control Testing

Evaluated by Western Blotting in PC12 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected GRP75 in PC12 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected GRP75 in COS-7, MCF-7, and NIH3T3 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected GRP75 in human kidney tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected GRP75 in MCF-7 cells.

Affinity Binding Assay: A representative lot of this antibody bound GRP75 peptide with a KD of 2.3 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Stress-70 protein, mitochondrial (UniProt: P38646; also known as 75 kDa glucose-regulated protein, GRP-75, Heat shock 70 kDa protein 9, Mortalin, MOT, Peptide-binding protein 74, PBP74) is encoded by the HSPA9 (also known as GRP75, HSPA9B, mt-HSP70) gene (Gene ID: 3313) in human. GRP75 is a mitochondrial resident protein that is synthesized with transit peptide (aa 1-46). It associates with the mitochondrial contact sites and cristae organizing system (MICOS) complex. It is a multipotent chaperone that regulates multiple cellular processes ranging from stress response to neurodegeneration. It plays a key role in several mitochondrial processes, including mitochondrial biogenesis, protein import, folding, and degradation. It has multiple binding partners, including p53, FGF-1, IL-1, IL-1R, HSP60, GRP94, and TIM and TOM. When associated with its binding partners, GRP75 has also reported to have various extramitochondrial functions, such as cell cycle arrest, apoptosis, senescence, and tumorigenesis. Dysregulation of GRP75-induced mitochondrial and proteasomal dysfunction has been suggested to be a contributor to the pathogenesis of Parkinson, Alzheimer s disease, diabetes mellitus, and cardiovascular diseases. Its levels are upregulated in multiple cancer types. Its interaction with p53 in the cytoplasm of transformed cells results in cytoplasmic retention and transcriptional inactivation of p53 leading to tumor growth. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Dhamad, AE., et al. (2020). Am. J. Physiol. 318(2); C289-C303; Wadhwa, R., et al. (2006). Int. J. Cancer. 118(12); 2973-2980).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service