Skip to Content
MilliporeSigma
All Photos(15)

Key Documents

ZRB1997

Sigma-Aldrich

Anti-EphA2 Antibody, clone 2C3 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Ephrin type-A receptor 2;EC: 2.7.10.1;Epithelial cell kinase;Tyrosine-protein kinase receptor ECK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C3, recombinant monoclonal

description

2C3 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 108.27 kDa
observed mol wt ~120 kDa

purified by

using Protein A

species reactivity

human, monkey

species reactivity (predicted by homology)

bovine, porcine, canine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... EPHA2(1969)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2C3 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Ephrin type-A receptor 2 (EphA2). It targets an epitope within 15 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Ephrin type-A receptor 2 (EphA2).

Application

Quality Control Testing

Evaluated by Western Blotting in PC3 cell lysates.

Western Blotting Analysis: 1:1,000 dilution of this antibody detected EphA2 in PC3 cell lysates.

Tested Applications

Western Blotting Analysis: 1:1,000 dilution from a representative lot detected EphA2 in A431, HEK293, and COS-7 cell lysates.

Affinity Binding Assay: A representative lot of this antibody bound EphA2 peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected EphA2 in human cerebral cortex tissue sections.

Flow Cytometry Analysis: 1 μg from a representative lot detected EphA2 in one million PC3 cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected EphA2 in A431 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Ephrin type-A receptor 2 (UniProt: P29317; also known as EC: 2.7.10.1, Epithelial cell kinase, Tyrosine-protein kinase receptor ECK) is encoded by the EPHA2 (also known as ECK) gene (Gene ID: 1969) in human. Ephrin type-A receptor 2 (EphA2) is a homodimeric single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-23), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 24-537), a short transmembrane domain (aa 538-558), and a cytoplasmic domain (aa 559-976). It is expressed in brain and glioma tissue and in glioma cell lines. It is also highly expressed in tissues that contain a high proportion of epithelial cells. Its levels are up-regulated by UV irradiation via a TP53-independent, MAPK-dependent mechanism. EphA2 serves as a receptor tyrosine kinase that binds promiscuously membrane-bound ephrin-A family ligands residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. It is involved in the regulation of migration, integrin-mediated adhesion, and in proliferation and differentiation of cells. EphA2 is phosphorylated on tyrosine residues upon binding and activation by EFNA1. Phosphorylation on tyrosine 588 and 594 is reported to be essential for binding VAV2 and VAV3 while phosphorylation on Tyr 735 and Tyr 930 are required for binding PI3-kinase p85 subunit. Overexpression of EphA2 is reported in murine models of skin tumors. Inflammatory cytokines are reported to promote RSK-dependent phosphorylation of EphA2 at serine 897 and this RSK-EphA2 axis might contribute to cell motility and promote tumor malignant progression. Two isoforms of Ephrin type-A receptor 2 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Zhou, Y., et al. (2015). Nat. Commun. 6; 7679; Guo, H., et al. (2006). Cancer Res. 66(14); 7050-7058).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service