Skip to Content
MilliporeSigma
All Photos(10)

Key Documents

ZRB1932

Sigma-Aldrich

Anti-TBKBP1 Antibody, clone 3F13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

TANK-binding kinase 1-binding protein 1;TBK1-binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3F13, recombinant monoclonal

description

3F13 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 67.7 kDa
observed mol wt ~75 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

mouse, porcine, canine, rat, bovine, feline

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TBKBP1(9755)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3F13 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects TANK-binding kinase 1-binding protein 1 (TBKBP1). It targets an epitope within 17 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human TANK-binding kinase 1-binding protein 1 (TBKBP1).

Application

Quality Control Testing

Evaluated by Western Blotting in NTERA-2 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected TBKBP1 in NTERA-2 cell lysate.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound TBKBP1 peptide with a KD of 2.1 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected TBKBP1 in human placenta tissue sections.

Western Blot Analysis: A 1:1000 dilution of a representative lot of the this antibody detected TBKBP1 in HEK293T cells transfected with a TBKBP1 expression vector.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

TANK-binding kinase 1-binding protein 1 (UniProt: A7MCY6; also known as TBK1-binding protein 1) is encoded by the TBKBP1 (also known as KIAA0775, SINTBAD) gene (Gene ID: 9755) in human. TBKBP1 is an adapter protein that constitutively binds TANK-binding kinase 1 (TBK1) and Inhibitor of NF-kB kinase subunit epsilon (IKBKE) and plays a role in antiviral innate immunity. Its TBK1 and IKBKE interacting region is localized in amino acids 280-329. It can exist as a homodimer or can form heterodimer with NAP1. Its homodimerization region is localized in amino acids 1-279. TBKBP1 expression is observed in leukocytes, lung, placenta, small intestine, liver, kidney, spleen, muscle, heart, and brain. TBK1 that is reported to be a central mediator of growth factor signaling relies on TBKBP1 for its action. TBKBP1 recruits TBK1 to protein kinase C (PKC ) through a scaffold protein, caspase recruitment domain 10 (CARD10). This enables PKC to phosphorylate TBK1 at serine 716. This phosphorylation is essential for TBK1 activation by growth factors, but not by innate immune stimuli. It has been reported that although the TBK1-TBKBP1 signaling axis is not essential for the induction of type I interferon, it facilitates mTORC1 activation and oncogenesis. Deletion of either TBK1 or TBKBP1 is known to inhibit tumorigenesis in a murine model of lung cancer. The TBK1-TBKBP1 axis can also facilitate tumor-mediated immunosuppression via induction of PD-L1 and stimulation of glycolysis. Two isoforms of TBKBP1 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Zhu, L., et al. (2019). Nat. Cell Biol. 21; 1604-1614; Goncalves, A., et al. (2011). PLoS One. 6(9); e23971).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service