Skip to Content
MilliporeSigma
All Photos(12)

Documents

ZRB1890

Sigma-Aldrich

Anti-QPCTL Antibody, clone 5M19 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:2.3.2.5, Glutaminyl-peptide cyclotransferase-like protein, Golgi-resident glutaminyl-peptide cyclotransferase, gQC, isoQC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5M19, recombinant monoclonal

description

5M19 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 42.92 kDa
observed mol wt ~38 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... QPCTL(54814)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 5M19 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Glutaminyl-peptide cyclotransferase-like protein (QCPTL). It targets an epitope within 15 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal half of human Glutaminyl-peptide cyclotransferase-like protein (QCPTL).

Application

Quality Control Testing

Evaluated by Western Blotting in Human liver microsomal preparations.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected QPCTL in Human liver microsomal preparations.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound QPCTL peptide with a KD of 5.8 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected QPCTL in Human stomach tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected QPCTL in HepG2 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Glutaminyl-peptide cyclotransferase-like protein (UniProt: Q9NXS2; also known as EC:2.3.2.5, Golgi-resident glutaminyl-peptide cyclotransferase, isoQC, gQC) is encoded by the QPCTL gene (Gene ID: 54814) in human. QPCTL is single-pass type I membrane protein that is exclusively localized within the Golgi complex. It contains a conserved zinc-binding motif (Asp-Glu-His), and its transmembrane domain is localized in amino acids 35-55. Its Golgi retention signal lies between amino acids 19 to 53. QPCTL is responsible for the biosynthesis of pryoglutamyl peptides. It catalyzes the cyclization of N-terminal glutamine and glutamic acid residues into an N-terminal pyroglutamate residue (pGlu).  It serves as a modulator of CD47-SIRPα binding. Inhibition of QPCTL is shown to enhance myeloid cell-mediated and antibody-dependent antitumor responses. It is also reported to regulate macrophage and monocyte abundance and inflammatory signatures in the tumor microenvironment. Inhibitors of QPCTL are reported to reduce pGlu-CD47 formation without affecting surface levels of CD47. Inhibition of pGlu formation is known to enhance the antitumor activity of myeloid cells similarly to antibody blockade of CD47 or SIRPα. Deletion of QPCTL is shown to control in vivo tumor growth. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Bresser, K., et al. (2022). Oncoimmunology. 11(1); 2049486; Logtenberg, MEW., et al. (2019). Nat. Med. 25(4); 612-619; Cynis, H., et al. (2008). J. Mol. Biol. 379(5); 966-980).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service