Skip to Content
MilliporeSigma
All Photos(18)

Key Documents

ZRB1698

Sigma-Aldrich

Anti-UBR5 Antibody, clone 2F14 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

E3 ubiquitin-protein ligase, E3 ubiquitin-protein ligase UBR5, EC:2.3.2.26, HECT domain-containing 1, HECT-type E3 ubiquitin transferase, Hyperplastic discs protein homolog, Progestin-induced protein, hHYD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

clone

recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 309.35 kDa
observed mol wt ~310 kDa

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2F14 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects E3 ubiquitin-protein ligase UBR5. It targets an epitope within the C-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 231 amino acids from the C-terminal half of human E3 ubiquitin-protein ligase UBR5.

Application

Quality Control Testing
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected UBR5 in HeLa cell lysate.Tested applicationsWestern Blotting Analysis: A 1:1,000 dilution from a representative lot detected UBR5 in NIH3T3, PC12 and HepG2 cell lysates.Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected UBR5 in HeLa, A431, and NIH 3T3 cells.Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected UBR5 in human testis tissue sections.Affinity Binding Assay: A representative lot of this antibody bound recombinant UBR5 protein fragment with a KD of 1.2 x 10-6 in an affinity binding assay.Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-UBR5, clone 2F14 ZooMAb, Cat. No. ZRB1698, is a recombinant Rabbit monoclonal antibody that specifically targets URB5 and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

E3 ubiquitin-protein ligase UBR5 (UniProt: O95071; also known as EC:2.3.2.26, E3 ubiquitin-protein ligase, HECT domain-containing 1, HECT-type E3 ubiquitin transferase, UBR5, Hyperplastic discs protein homolog, hHYD, Progestin-induced protein) is encoded by the URB5 (also known as EDD, EDD1, HYD, KIAA0896) gene (Gene ID: 51366) in human. UBR5 is a widely expressed nuclear protein that is shown to be most abundant in testis. It is also expressed at high levels in brain, pituitary, and kidney. URB5 is identified as a novel regulator of p53 and downregulation of UBR5 results in elevated levels of p53 in normal and transformed cells. Its deficiency in mice is reported to result in mid gestation embryonic death due to a compromised yolk sac vascular development. UBR5 is a component of the N-end rule pathway. It recognizes and binds to proteins bearing specific N-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation. It is shown to be involved in maturation and/or transcriptional regulation of mRNA by activating CDK9 by polyubiquitination. It also plays a role in the control of cell cycle progression. It associates with CDK9 and TFIIS/TCEA1 and forms a transcription regulatory complex made of CDK9, RNAP II, UBR5, and TFIIS/TCEA1 that can stimulate target gene transcription. UBR5 regulates DNA topoisomerase II binding protein (TopBP1) in the DNA damage response and may have a role in tumor suppression. Two isoforms of UBR5 are described that are produced by alternative splicing and differ only in a single amino acid. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Smits., VAJ. (2012). Cell Cycle 11(4); 715-720).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service