Skip to Content
MilliporeSigma
All Photos(7)

Documents

ZRB1669

Sigma-Aldrich

Anti-p-RPS6-Ser235/236 Antibody, clone 1I22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

40S ribosomal protein S6;Phosphoprotein NP33;Small ribosomal subunit protein eS6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1I22, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 28.68 kDa
observed mol wt ~33 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

bovine, sheep, mouse, rat, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer235/pSer236)

Gene Information

human ... RPS6(6194)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1I22 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects 40S ribosomal protein S6 phosphorylated on serine 235/236.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphoserines 235/236 from the C-terminal region of human 40S ribosomal protein S6.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HeLa cells treated with Calyculin A and Okadaic Acid.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected phospho-S6 Ribosomal protein (RPS6)-Ser235/236 in lysate from HeLa cells overnight serum starved and treated with Calyculin A (50 nM) and Okadaic acid (500 nM) for 30 min.

Tested applications

Affinity Binding Assay: A representative lot of this antibody bound p-RPS6-Ser235/236 peptide with a KD of 8.7 x 10-6 in an affinity binding assay.

Peptide Inhibition Assay Analysis: : Target band detection in HeLa cell lysate was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in lysate from HeLa cells treated with Calyculin A and Okadaic Acid.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected phospho-S6 Ribosomal protein (RPS6)-Ser235/236 in lysate from HeLa cells overnight serum starved and treated with Calyculin A (50 nM) and Okadaic acid (500 nM) for 30 min.

Target description

40S ribosomal protein S6 (UniProt: P62753; also known as Phosphoprotein NP33, Small ribosomal subunit protein eS6) is encoded by the RPS6 (also known as OK/SW-cl.2) gene (Gene ID: 6194) in human. RPS6 is a highly conserved member of the eukaryotic ribosomal protein S6 family and is a component of the 40S ribosomal subunit. It plays an important role in controlling cell growth and proliferation through the selective translation of particular classes of mRNA. It is the major substrate of protein kinases in eukaryote ribosomes and its phosphorylation is stimulated by growth factors, tumor promoting agents, and mitogens. RPS6 can be phosphorylated on five serine residues (Ser235, 236, 240, 244, and 247) in the region close to the carboxy terminus. Among the serine residues 235 and 236 are shown to be directly phosphorylated by S6 Kinase that is subjected to regulation by the nutrient-mTORC1 signaling. Phosphorylation at serine 240/244 is reported to be predominantly nuclear but is also detectable in the cytoplasm, whereas serine 235/236 phosphorylated form is almost exclusively localized to the nucleus of primary human cells and is virtually undetectable in the cytoplasm, except in transformed cells where it can also be detected in the cytoplasm. RPS6 is also shown to exist in complex with the Kaposi s sarcoma-associated herpesvirus latency-associated nuclear antigen (LANA). This interaction is mediated by the N-terminal domain of LANA. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Rosner, M., et al. (2011). Amino Acids 40(2); 595-600; Chen, W., and Dittmar, DP. (2011). J. Virol. 85(18); 9495-9505; Biever, A., et al. (2015). J. Neurosci. 2015, 35(10); 4113-4130).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service